Gene Transcripts

Gene Symbol
Rrp9
Entrez Gene ID
363134
Gene Name
ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
Rnu3ip2
Chromosome Location
Chr. 8: 115179899 - 115188551 on Build Rnor_6.0
UniGene ID
Rn.11840
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108778.1 NP_001102248.1 6, 7 567
GenBank mRNA BC158778.1 - 6, 7 564
FQ230405.1 - 6, 7 550

View Full Product Details