Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135760.1 | NP_001129232.1 | 10 | 1670 | |
XM_006248639.2 | XP_006248701.1 | 11 | 971 | ||
XM_006248641.2 | XP_006248703.1 | 8 | 1050 | ||
XM_008768893.1 | XP_008767115.1 | 10 | 3473 | ||
XM_008768894.1 | XP_008767116.1 | 10 | 1663 | ||
XM_008768895.1 | XP_008767117.1 | 10 | 1780 | ||
XM_008768896.1 | XP_008767118.1 | 9 | 1430 | ||
XM_008768897.1 | XP_008767119.1 | 11 | 1370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008768905.1 | XP_008767127.1 | 2957 | ||
XM_008768906.1 | XP_008767128.1 | 1501 | |||
XM_008768907.1 | XP_008767129.1 | 1430 | |||
XM_008768908.1 | XP_008767130.1 | 2028 | |||
XM_008768909.1 | XP_008767131.1 | 972 | |||
XM_008768910.1 | XP_008767132.1 | 1050 |