Gene Transcripts

Gene Symbol
Reep5
Entrez Gene ID
364838
Gene Name
receptor accessory protein 5
Gene Aliases
Dp1
Chromosome Location
Chr. 18: 27128462 - 27159693 on Build Rnor_6.0
UniGene ID
Rn.40477
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108888.2 NP_001102358.2 2 276
GenBank mRNA BC167013.1 - 2 201
BE126660.1 - 2 276
FQ213789.1 - 2 208
FQ235123.1 - 2 208

View Full Product Details