Gene Transcripts

Gene Symbol
Syce2
Entrez Gene ID
364976
Gene Name
synaptonemal complex central element protein 2
Gene Aliases
RGD1311867
Chromosome Location
Chr. 19: 25981601 - 26002399 on Build Rnor_6.0
UniGene ID
Rn.230993
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191557.1 NP_001178486.1 2, 3 308
XM_006255291.2 XP_006255353.1 3, 4 1858

View Full Product Details