Gene Transcripts

Gene Symbol
Uqcrh
Entrez Gene ID
366448
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases
Chromosome Location
Chr. 5: 134919040 - 134927235 on Build Rnor_6.0
UniGene ID
Rn.7401
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009480.1 NP_001009480.1 3 202
GenBank mRNA BC086954.1 - 3 202
FQ217127.1 - 3 247
FQ217158.1 - 3 242
FQ217230.1 - 3 239
FQ217709.1 - 3 246
FQ217710.1 - 3 232
FQ217725.1 - 3 243
FQ218028.1 - 3 232
FQ221521.1 - 3 246
FQ221648.1 - 3 246
FQ221867.1 - 3 235
FQ222083.1 - 3 239
FQ223801.1 - 3 232
FQ223893.1 - 3 246
FQ223943.1 - 3 243
FQ224007.1 - 3 244
FQ224711.1 - 3 217

View Full Product Details