Gene Transcripts

Gene Symbol
Tcf20
Entrez Gene ID
366964
Gene Name
transcription factor 20
Gene Aliases
AR1, SPBP
Chromosome Location
Chr. 7: 123670135 - 123767797 on Build Rnor_6.0
UniGene ID
Rn.213688
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130574.1 NP_001124046.1 2 4544
XM_006242110.2 XP_006242172.1 1 5444
XM_006242112.2 XP_006242174.1 1 5444
XM_008765772.1 XP_008763994.1 1 5443

View Full Product Details