Gene Transcripts
- Gene Symbol
- Mcpt8l3
- Entrez Gene ID
- 408207
- Gene Name
- mast cell protease 8-like 3
- Gene Aliases
- Chromosome Location
- Chr. 15: 34667042 - 34670290 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_573781.6 |
XP_573781.1
|
2 |
205 |
|
View Full Product Details