Gene Transcripts

Gene Symbol
Ly49i3
Entrez Gene ID
494208
Gene Name
immunoreceptor Ly49i3
Gene Aliases
Chromosome Location
Chr. 4: 164743490 - 164747388 on Build Rnor_6.0
UniGene ID
Rn.55469
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009499.1 NP_001009499.1 218
XM_008763437.1 XP_008761659.1 1 527
XM_008763438.1 XP_008761660.1 2 485
GenBank mRNA AY659932.1 - 218

Gene Symbol
Ly49i4
Entrez Gene ID
494204
Gene Name
Ly49 inhibitory receptor 4
Gene Aliases
Chromosome Location
Chr. 4: 164515105 - 164536581 on Build Rnor_6.0
UniGene ID
Rn.83631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009495.1 NP_001009495.1 3 305
XM_008763405.1 XP_008761627.1 4 420
GenBank mRNA AY649834.1 - 3 305

View Full Product Details