Gene Transcripts
- Gene Symbol
- ZMYM4
- Entrez Gene ID
- 9202
- Gene Name
- zinc finger MYM-type containing 4
- Gene Aliases
- CDIR, MYM, ZNF198L3, ZNF262
- Chromosome Location
- Chr. 1: 35268694 - 35421944 on Build GRCh38
- UniGene ID
- Hs.269211
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005095.2 |
NP_005086.2
|
17, 18 |
2848 |
|
XM_005271328.4 |
XP_005271385.1
|
17, 18 |
3127 |
|
XM_005271331.2 |
XP_005271388.1
|
14, 15 |
2093 |
|
XM_011542421.2 |
XP_011540723.1
|
17, 18 |
2774 |
|
XM_011542423.2 |
XP_011540725.1
|
18, 19 |
3179 |
|
XM_011542425.2 |
XP_011540727.1
|
15, 16 |
2133 |
|
XM_017002803.1 |
XP_016858292.1
|
18, 19 |
2901 |
|
XM_017002804.1 |
XP_016858293.1
|
18, 19 |
2841 |
|
XM_017002805.1 |
XP_016858294.1
|
13, 14 |
2339 |
|
GenBank mRNA |
AB007885.1 |
- |
13, 14 |
1968 |
|
AK309705.1 |
- |
16, 17 |
2508 |
|
BC012093.1 |
- |
15, 16 |
2427 |
|
BC127113.1 |
- |
18, 19 |
2854 |
40125110 |
BC127114.1 |
- |
14, 15 |
2037 |
40125115 |
BX647376.1 |
- |
13, 14 |
2079 |
|
BX648067.1 |
- |
8, 9 |
1142 |
|
CR936741.1 |
- |
16, 17 |
2712 |
|
View Full Product Details