Gene Transcripts

Gene Symbol
Prss36
Entrez Gene ID
497040
Gene Name
protease, serine, 36
Gene Aliases
Chromosome Location
Chr. 1: 199379374 - 199396217 on Build Rnor_6.0
UniGene ID
Rn.57253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011560.2 NP_001011560.2 12 1811
XM_008759913.1 XP_008758135.1 12 2698

View Full Product Details