Gene Transcripts

Gene Symbol
Mrps24
Entrez Gene ID
498406
Gene Name
mitochondrial ribosomal protein S24
Gene Aliases
DD6A4-3
Chromosome Location
Chr. 14: 85947763 - 85951891 on Build Rnor_6.0
UniGene ID
Rn.1914
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077659.1 NP_001071127.1 3 190
GenBank mRNA BC157807.1 - 3 186
BP503351.1 - 3 195
DY319747.1 - 3 186
FQ227686.1 - 3 208

View Full Product Details