Gene Transcripts

Gene Symbol
Spata2L
Entrez Gene ID
498963
Gene Name
spermatogenesis associated 2-like
Gene Aliases
RGD1560840
Chromosome Location
Chr. 19: 56032607 - 56037850 on Build Rnor_6.0
UniGene ID
Rn.91547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109133.1 NP_001102603.1 2 1107
XM_006255781.2 XP_006255843.1 3 1560

View Full Product Details