Gene Transcripts

Gene Symbol
Cbln4
Entrez Gene ID
499947
Gene Name
cerebellin 4 precursor
Gene Aliases
Chromosome Location
Chr. 3: 170033166 - 170040953 on Build Rnor_6.0
UniGene ID
Rn.75276
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109210.1 NP_001102680.1 3 1691

View Full Product Details