Gene Transcripts

Gene Symbol
Doxl2
Entrez Gene ID
312317
Gene Name
diamine oxidase-like protein 2
Gene Aliases
Chromosome Location
Chr. 4: 78618743 - 78622525 on Build Rnor_6.0
UniGene ID
Rn.213049
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199291.1 NP_954985.1 1 386
XM_006236462.1 XP_006236524.1 2 412
GenBank mRNA BC098900.1 - 2 418

View Full Product Details