Gene Transcripts

Gene Symbol
Eqtn
Entrez Gene ID
500502
Gene Name
equatorin, sperm acrosome associated
Gene Aliases
Afaf
Chromosome Location
Chr. 5: 113852191 - 113881098 on Build Rnor_6.0
UniGene ID
Rn.126103
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039345.1 NP_001034434.1 3 384
XM_008763895.1 XP_008762117.1 3 571
XM_008763896.1 XP_008762118.1 3 571
GenBank mRNA DQ336136.1 - 3 384

View Full Product Details