Gene Transcripts

Gene Symbol
Timm17al1
Entrez Gene ID
500655
Gene Name
translocase of inner mitochondrial membrane 17a-like 1
Gene Aliases
RGD1560665
Chromosome Location
Chr. 6: 83252019 - 83253099 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_576033.5 XP_576033.1 1 77

View Full Product Details