Gene Transcripts

Gene Symbol
Plscr5
Entrez Gene ID
501036
Gene Name
phospholipid scramblase family, member 5
Gene Aliases
RGD1564500
Chromosome Location
Chr. 8: 99558395 - 99579336 on Build Rnor_6.0
UniGene ID
Rn.228189
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191999.1 NP_001178928.1 3 299
XM_006243540.2 XP_006243602.1 5 634

View Full Product Details