Gene Transcripts

Gene Symbol
FADS2
Entrez Gene ID
9415
Gene Name
fatty acid desaturase 2
Gene Aliases
D6D, DES6, FADSD6, LLCDL2, SLL0262, TU13
Chromosome Location
Chr. 11: 61816203 - 61867354 on Build GRCh38
UniGene ID
Hs.502745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281501.1 NP_001268430.1 6 812
NM_001281502.1 NP_001268431.1 6 881
NM_004265.3 NP_004256.1 6 1153
GenBank mRNA AF084559.1 - 6 945
AF108658.1 - 6 738
AF126799.1 - 6 808
AK074925.1 - 4 360
AK074939.1 - 6 885
AK074991.1 - 6 621
AK123240.1 - 1 782
AK290291.1 - 6 759
AK299762.1 - 6 881
AL050118.1 - 4 397
BC009011.1 - 6 887 4181037

View Full Product Details