Gene Transcripts

Gene Symbol
Nobox
Entrez Gene ID
502759
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
RGD1563929
Chromosome Location
Chr. 4: 72749478 - 72757066 on Build Rnor_6.0
UniGene ID
Rn.228183
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001192013.1 NP_001178942.1 8 989
XM_008762912.1 XP_008761134.1 8 1184

View Full Product Details