Gene Transcripts

Gene Symbol
Vom2r8
Entrez Gene ID
685641
Gene Name
vomeronasal 2 receptor, 8
Gene Aliases
Chromosome Location
Chr. Un|NW_007906786: 18673 - 58717 on Build Rnor_6.0
UniGene ID
Rn.201688
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099464.1 NP_001092934.1 1097

View Full Product Details