Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190411.1 | NP_001177340.1 | 4 | 495 | |
NM_001190412.1 | NP_001177341.1 | 5 | 671 | ||
NM_001190413.1 | NP_001177342.1 | 4 | 495 | ||
NM_006987.3 | NP_008918.1 | 5 | 671 | ||
XM_011535364.2 | XP_011533666.1 | 4 | 540 | ||
XM_011535365.2 | XP_011533667.1 | 4 | 480 | ||
XM_011535366.2 | XP_011533668.1 | 4 | 422 | ||
XM_011535367.2 | XP_011533669.1 | 4 | 422 | ||
XM_011535368.2 | XP_011533670.1 | 4 | 468 | ||
XM_011535369.2 | XP_011533671.1 | 4 | 403 | ||
XM_017025367.1 | XP_016880856.1 | 4 | 540 | ||
XM_017025368.1 | XP_016880857.1 | 4 | 540 | ||
XM_017025369.1 | XP_016880858.1 | 4 | 430 | ||
XM_017025370.1 | XP_016880859.1 | 6 | 979 | ||
XM_017025371.1 | XP_016880860.1 | 4 | 540 | ||
GenBank mRNA | AF129812.1 | - | 4 | 576 | |
AK000469.1 | - | 5 | 614 | ||
AK291950.1 | - | 4 | 464 | ||
BC005153.1 | - | 5 | 935 | 3529317 | |
BC093776.1 | - | 3 | 365 | 7939621 | |
BC113413.1 | - | 3 | 365 | 8322465 | |
BU197069.1 | - | 4 | 438 | ||
DA378956.1 | - | 4 | 495 |