Gene Transcripts

Gene Symbol
Upk3b
Entrez Gene ID
360790
Gene Name
uroplakin 3B
Gene Aliases
RGD1309615
Chromosome Location
Chr. 12: 23676169 - 23682394 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249208.2 XP_006249270.1 1, 2 266
XM_341056.7 XP_341057.3 1, 2 266

View Full Product Details