Gene Transcripts

Gene Symbol
Cnpy1
Entrez Gene ID
685001
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
Chromosome Location
Chr. 4: 474424 - 483668 on Build Rnor_6.0
UniGene ID
Rn.233005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749650.3 XP_003749698.1 2 898
GenBank mRNA AB190508.1 - 1 91

View Full Product Details