Gene Transcripts
- Gene Symbol
- MDC1
- Entrez Gene ID
- 9656
- Gene Name
- mediator of DNA damage checkpoint 1
- Gene Aliases
- NFBD1
- Chromosome Location
- Chr. 6: 30699807 - 30717988 on Build GRCh38
- UniGene ID
- Hs.653495
- Species
- Homo sapiens
- Gene Symbol
- MDC1-AS1
- Entrez Gene ID
- 106478956
- Gene Name
- MDC1 antisense RNA 1
- Gene Aliases
- MDC1-AS
- Chromosome Location
- Chr. 6: 30703067 - 30713184 on Build GRCh38
- UniGene ID
- Hs.708954
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BQ575709.1 |
- |
1 |
473 |
|
View Full Product Details