Gene Transcripts

Gene Symbol
Ccdc84
Entrez Gene ID
689046
Gene Name
coiled-coil domain containing 84
Gene Aliases
Chromosome Location
Chr. 8: 48729695 - 48736579 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001060013.5 XP_001060013.1 8 771
XM_008766177.1 XP_008764399.1 7 748
XM_008766178.1 XP_008764400.1 8 792

View Full Product Details