Gene Transcripts

Gene Symbol
Fam83c
Entrez Gene ID
683506
Gene Name
family with sequence similarity 83, member C
Gene Aliases
Chromosome Location
Chr. 3: 151365675 - 151371567 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749599.3 XP_003749647.1 4 2191

View Full Product Details