Gene Transcripts

Gene Symbol
Catsper4
Entrez Gene ID
362623
Gene Name
cation channel, sperm associated 4
Gene Aliases
RGD1565213
Chromosome Location
Chr. 5: 152430048 - 152446808 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_342941.7 XP_342942.5 7 855

View Full Product Details