Gene Transcripts

Gene Symbol
Uqcr10
Entrez Gene ID
685322
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Gene Aliases
Ucrc
Chromosome Location
Chr. 14: 84935480 - 84937741 on Build Rnor_6.0
UniGene ID
Rn.140706
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170465.1 NP_001163936.1 2 357
GenBank mRNA BC166935.1 - 2 341

View Full Product Details