Gene Transcripts

Gene Symbol
Rnf11l2
Entrez Gene ID
316552
Gene Name
ring finger protein 11-like 2
Gene Aliases
Rnf11, Rnf11l
Chromosome Location
Chr. 9: 86652248 - 86654050 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245310.2 XP_006245372.1 1 270

View Full Product Details