Gene Transcripts

Gene Symbol
Aknad1
Entrez Gene ID
691416
Gene Name
AKNA domain containing 1
Gene Aliases
Chromosome Location
Chr. 2: 211541828 - 211585332 on Build Rnor_6.0
UniGene ID
Rn.153981
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044303.2 NP_001037768.2 5, 6 1414
XM_008761422.1 XP_008759644.1 5, 6 2595
GenBank mRNA BC079051.1 - 5, 6 1414

View Full Product Details