Gene Transcripts

Gene Symbol
Snrnp27
Entrez Gene ID
362392
Gene Name
small nuclear ribonucleoprotein 27 (U4/U6.U5)
Gene Aliases
RGD1310925, Ry1
Chromosome Location
Chr. 4: 118478075 - 118487633 on Build Rnor_6.0
UniGene ID
Rn.19010
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108636.1 NP_001102106.1 5, 6 456
GenBank mRNA FQ224589.1 - 458

View Full Product Details