Gene Transcripts

Gene Symbol
Fam53c
Entrez Gene ID
685405
Gene Name
family with sequence similarity 53, member C
Gene Aliases
Chromosome Location
Chr. 18: 27558091 - 27572633 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001062862.5 XP_001062862.1 4 440
XM_006254613.2 XP_006254675.1 4 621
XM_008772055.1 XP_008770277.1 3 382

View Full Product Details