Gene Transcripts

Gene Symbol
Srrm3
Entrez Gene ID
685890
Gene Name
serine/arginine repetitive matrix 3
Gene Aliases
RGD1307391
Chromosome Location
Chr. 12: 23855400 - 23925741 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277394.1 NP_001264323.1 9, 10 874

View Full Product Details