Gene Transcripts

Gene Symbol
Rpl38
Entrez Gene ID
689284
Gene Name
ribosomal protein L38
Gene Aliases
Chromosome Location
Chr. 10: 103198424 - 103202059 on Build Rnor_6.0
UniGene ID
Rn.129626
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077592.1 NP_001071060.1 4 175
XM_008768393.1 XP_008766615.1 4 174
GenBank mRNA BC101933.1 - 4 175
FQ210073.1 - 4 182
FQ217659.1 - 4 177
FQ222790.1 - 4 182
FQ223953.1 - 4 182
FQ224522.1 - 4 182
FQ228832.1 - 4 193
X57007.1 - 4 180

Gene Symbol
LOC685963
Entrez Gene ID
685963
Gene Name
similar to 60S ribosomal protein L38
Gene Aliases
Chromosome Location
Chr. 3: 164002409 - 164002757 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001065954.5 XP_001065954.1 1 197

Gene Symbol
LOC690468
Entrez Gene ID
690468
Gene Name
similar to 60S ribosomal protein L38
Gene Aliases
Chromosome Location
Chr. 2: 199478992 - 199479315 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001074532.4 XP_001074532.1 1 179

View Full Product Details