Gene Transcripts

Gene Symbol
Msgn1
Entrez Gene ID
689864
Gene Name
mesogenin 1
Gene Aliases
Chromosome Location
Chr. 6: 36876165 - 36876805 on Build Rnor_6.0
UniGene ID
Rn.21210
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109551.1 NP_001103021.1 1 496

View Full Product Details