Gene Transcripts

Gene Symbol
Amtn
Entrez Gene ID
689404
Gene Name
amelotin
Gene Aliases
Chromosome Location
Chr. 14: 21286509 - 21300620 on Build Rnor_6.0
UniGene ID
Rn.154394
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044296.1 NP_001037761.1 3 207
XM_008770057.1 XP_008768279.1 2 1863
GenBank mRNA AM231717.1 - 3 167
DQ198381.1 - 3 207

View Full Product Details