Gene Transcripts

Gene Symbol
Rps4y2
Entrez Gene ID
690845
Gene Name
ribosomal protein S4, Y-linked 2
Gene Aliases
Chromosome Location
Chr. 4: 182745448 - 182746381 on Build Rnor_6.0
UniGene ID
Rn.107176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109612.1 NP_001103082.1 1 540

View Full Product Details