Gene Transcripts

Gene Symbol
Vps37d
Entrez Gene ID
687208
Gene Name
vacuolar protein sorting 37 homolog D (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 12: 24651342 - 24656793 on Build Rnor_6.0
UniGene ID
Rn.217242
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128193.2 NP_001121665.1 4 1415
GenBank mRNA BC166560.1 - 4 1415

View Full Product Details