Gene Transcripts

Gene Symbol
PLEKHM1
Entrez Gene ID
9842
Gene Name
pleckstrin homology and RUN domain containing M1
Gene Aliases
AP162, B2, OPTB6
Chromosome Location
Chr. 17: 45434262 - 45490780 on Build GRCh38
UniGene ID
Hs.514242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014798.2 NP_055613.1 8 2787
NR_027774.1 7 2539
NR_027782.1 7 2402
XM_006722201.3 XP_006722264.1 6 2568
XM_011525523.2 XP_011523825.1 8 2758
XM_011525524.1 XP_011523826.1 8 2937
XM_011525525.1 XP_011523827.1 7 2545
XM_011525526.1 XP_011523828.1 9 2581
XM_011525528.2 XP_011523830.1 3 1131
XM_017025451.1 XP_016880940.1 8 2735
XM_017025452.1 XP_016880941.1 7 2490
XM_017025453.1 XP_016880942.1 9 2944
XM_017025454.1 XP_016880943.1 7 2635
GenBank mRNA AB002354.2 - 8 2740
AJ002220.1 - 6 2375
AK074103.1 - 4 1535
AK299116.1 - 7 2487
AK299467.1 - 7 2541
BC064361.1 - 8 2751 5535600

Gene Symbol
PLEKHM1P1
Entrez Gene ID
440456
Gene Name
pleckstrin homology and RUN domain containing M1 pseudogene 1
Gene Aliases
PLEKHM1P
Chromosome Location
Chr. 17: 64784841 - 64837184 on Build GRCh38
UniGene ID
Hs.646585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024386.2 7 1953
GenBank mRNA AL833315.1 - 7 1923

View Full Product Details