Gene Transcripts

Gene Symbol
Spem1
Entrez Gene ID
691981
Gene Name
spermatid maturation 1
Gene Aliases
Chromosome Location
Chr. 10: 56428447 - 56429748 on Build Rnor_6.0
UniGene ID
Rn.60878
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109653.1 NP_001103123.1 1 193

View Full Product Details