Gene Transcripts

Gene Symbol
Slc16a5
Entrez Gene ID
690212
Gene Name
solute carrier family 16 (monocarboxylate transporter), member 5
Gene Aliases
Chromosome Location
Chr. 10: 104019888 - 104035725 on Build Rnor_6.0
UniGene ID
Rn.198898
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109568.1 NP_001103038.1 4 1420
XM_006247735.2 XP_006247797.1 4 1234
XM_008768396.1 XP_008766618.1 4 1394

View Full Product Details