Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005110.3 | NP_005101.1 | 15 | 1664 | |
XM_006714942.2 | XP_006715005.1 | 15 | 1613 | ||
XM_017010100.1 | XP_016865589.1 | 14 | 1549 | ||
GenBank mRNA | AB016789.1 | - | 15 | 1593 | |
AK001242.1 | - | 5 | 546 | ||
AK022507.1 | - | 15 | 1604 | ||
AK297750.1 | - | 13 | 1300 | ||
AK314551.1 | - | 15 | 1604 | ||
BC000012.2 | - | 15 | 1602 | 3506723 | |
BT009818.1 | - | 14 | 1494 | ||
KU178494.1 | - | 14 | 1494 | ||
KU178495.1 | - | 14 | 1494 |