Gene Transcripts

Gene Symbol
Ssx1
Entrez Gene ID
691903
Gene Name
synovial sarcoma, X breakpoint 1
Gene Aliases
Chromosome Location
Chr. X: 14903854 - 14912000 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773085.1 XP_008771307.1 5 1613

View Full Product Details