Gene Transcripts

Gene Symbol
Gp1ba
Entrez Gene ID
691992
Gene Name
glycoprotein Ib (platelet), alpha polypeptide
Gene Aliases
Chromosome Location
Chr. 10: 57260680 - 57263546 on Build Rnor_6.0
UniGene ID
Rn.218581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109654.1 NP_001103124.1 2 1008

View Full Product Details