Gene Transcripts

Gene Symbol
HBM
Entrez Gene ID
3042
Gene Name
hemoglobin subunit mu
Gene Aliases
HBAP2, HBK
Chromosome Location
Chr. 16: 165974 - 166768 on Build GRCh38
UniGene ID
Hs.647389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003938.3 NP_001003938.1 3 440
GenBank mRNA AY698022.2 - 3 440
BC035682.1 - 3 423 5749846

View Full Product Details