Gene Transcripts

Gene Symbol
SH2B3
Entrez Gene ID
10019
Gene Name
SH2B adaptor protein 3
Gene Aliases
IDDM20, LNK
Chromosome Location
Chr. 12: 111405106 - 111451623 on Build GRCh38
UniGene ID
Hs.506784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291424.1 NP_001278353.1 2 249
NM_005475.2 NP_005466.1 3 1172
XM_005253818.4 XP_005253875.1 4 1316
XM_005253819.4 XP_005253876.1 3 1196
XM_006719180.3 XP_006719243.1 2 798
XM_011537719.2 XP_011536021.1 4 1316
XM_011537720.2 XP_011536022.1 4 981
XM_011537721.2 XP_011536023.1 2 2781
GenBank mRNA AB208911.1 - 2 408
AF055581.1 - 3 1172
AJ012793.1 - 2 815
AK302173.1 - 2 249
BC136451.1 - 3 858 9020440

View Full Product Details