Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001258450.1 | NP_001245379.1 | 4 | 719 | |
NM_001258451.1 | NP_001245380.1 | 4 | 790 | ||
NM_004203.4 | NP_004194.3 | 4 | 926 | ||
NM_182687.2 | NP_872629.1 | 4 | 926 | ||
XM_011522734.2 | XP_011521036.1 | 4 | 887 | ||
XM_011522735.2 | XP_011521037.1 | 4 | 713 | ||
XM_011522736.2 | XP_011521038.1 | 3 | 720 | ||
GenBank mRNA | AK097642.1 | - | 4 | 675 | |
AK098452.1 | - | 4 | 778 | ||
AK294379.1 | - | 3 | 687 | ||
AK301926.1 | - | 4 | 595 | ||
AK303005.1 | - | 4 | 804 | ||
AK309964.1 | - | 3 | 720 | ||
BC121161.1 | - | 3 | 614 | ||
BC121162.1 | - | 3 | 614 | ||
BC130011.1 | - | 3 | 614 | ||
BC153823.1 | - | 3 | 614 | ||
U56816.1 | - | 3 | 630 |