Gene Transcripts
- Gene Symbol
- TMSB10
- Entrez Gene ID
- 9168
- Gene Name
- thymosin beta 10
- Gene Aliases
- MIG12, TB10
- Chromosome Location
- Chr. 2: 84905639 - 84906675 on Build GRCh38
- UniGene ID
- Hs.446574
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021103.3 |
NP_066926.1
|
2, 3 |
192 |
|
GenBank mRNA |
AK130949.1 |
- |
2, 3 |
180 |
|
AK311952.1 |
- |
2, 3 |
175 |
|
AY453400.1 |
- |
1, 2 |
125 |
|
BC016025.1 |
- |
2, 3 |
157 |
3909431 |
BC016731.1 |
- |
2, 3 |
179 |
4095378 |
BC107889.1 |
- |
2, 3 |
179 |
|
M20259.1 |
- |
2, 3 |
157 |
|
M92381.1 |
- |
1, 2 |
114 |
|
S54005.1 |
- |
2, 3 |
162 |
|
View Full Product Details