Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052863.2 | NP_443095.2 | 1, 2 | 88 | |
GenBank mRNA | AF086152.1 | - | 1, 2 | 84 | |
AF313458.1 | - | 1, 2 | 73 | ||
AF436839.1 | - | 1, 2 | 84 | ||
AY359064.1 | - | 1, 2 | 141 | ||
BC029176.1 | - | 1, 2 | 78 | 5180304 | |
BC072673.1 | - | 1, 2 | 84 | 5753142 |