Gene Transcripts

Gene Symbol
NDUFA11
Entrez Gene ID
126328
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases
B14.7, CI-B14.7
Chromosome Location
Chr. 19: 5891276 - 5904013 on Build GRCh38
UniGene ID
Hs.406062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175614.4 NP_783313.1 4 771
NR_034166.2 5 891
GenBank mRNA AL546175.3 - 5 631
AY726561.1 - 3 2125
BC069045.1 - 4 526 6503302
BQ054257.1 - 4 527
BQ671956.1 - 4 545

View Full Product Details